Assay Details
| Gene Symbol: sort1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213230.1 | NP_998395.1 | 16 - 17 | 2075 | 72 | |
| GenBank mRNA | BC068433.1 | - | 16 - 17 | 2075 | 72 | |
More Information
Set Membership: |
|

sort1a
Genomic Map