Assay Details
| Gene Symbol: sort1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213230.1 | NP_998395.1 | 2 - 3 | 364 | 88 | |
| XM_005172902.4 | XP_005172959.1 | 1 - 2 | 413 | 88 | ||
| GenBank mRNA | BC068433.1 | - | 2 - 3 | 364 | 88 | |
More Information
Set Membership: |
|

sort1a
Genomic Map