Assay Details
| Gene Symbol: sort1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213230.1 | NP_998395.1 | 9 - 10 | 1014 | 80 | |
| XM_005172902.4 | XP_005172959.1 | 8 - 9 | 1063 | 80 | ||
| GenBank mRNA | BC068433.1 | - | 9 - 10 | 1014 | 80 | |
More Information
Set Membership: |
|

sort1a
Genomic Map