Assay Details
| Gene Symbol: prpf38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213489.1 | NP_998654.1 | 3 - 4 | 645 | 70 | |
| XM_005171371.4 | XP_005171428.1 | 3 - 4 | 675 | 70 | ||
| XM_005171372.4 | XP_005171429.1 | 3 - 4 | 675 | 70 | ||
| XM_005171373.1 | XP_005171430.1 | 2 - 3 | 174 | 70 | ||
| XM_009298770.3 | XP_009297045.1 | 3 - 4 | 666 | 70 | ||
| XM_009298771.2 | XP_009297046.1 | 3 - 4 | 280 | 70 | ||
| GenBank mRNA | BC061448.1 | - | 3 - 4 | 645 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005171372.4, XM_005171373.1, XM_009298770.3, XM_009298771.2 transcripts.
Set Membership: |
|

prpf38b
Genomic Map