Assay Details
| Gene Symbol: prkcda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214708.1 | NP_999873.1 | 4 - 5 | 468 | 104 | |
| GenBank mRNA | BC049327.1 | - | 4 - 5 | 468 | 104 | |
More Information
Set Membership: |
|

prkcda
Genomic Map