Assay Details
| Gene Symbol: mob1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214783.2 | NP_999948.2 | 4 - 5 | 474 | 65 | |
| GenBank mRNA | AY398317.1 | - | 4 - 5 | 470 | 65 | |
| BC095569.1 | - | 4 - 5 | 474 | 65 | ||
More Information
Set Membership: |
|

mob1bb
Genomic Map