Assay Details
| Gene Symbol: asic1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214789.1 | NP_999954.1 | 7 - 8 | 1307 | 147 | |
| XM_017353815.2 | XP_017209304.1 | 7 - 8 | 1500 | 147 | ||
| XM_021470127.1 | XP_021325802.1 | 7 - 8 | 1504 | 147 | ||
| XM_021470128.1 | XP_021325803.1 | 7 - 8 | 1507 | 147 | ||
| XM_021470129.1 | XP_021325804.1 | 7 - 8 | 2061 | 147 | ||
| XM_021470130.1 | XP_021325805.1 | 7 - 8 | 1503 | 147 | ||
| GenBank mRNA | AJ609617.1 | - | 7 - 8 | 1307 | 147 | |
More Information
Set Membership: |
|

asic1c
Genomic Map