Assay Details
| Gene Symbol: epb41l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214812.1 | NP_999977.1 | 1165 | 77 | ||
| GenBank mRNA | BC070025.1 | - | 1165 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

epb41l3b| Gene Symbol: epb41l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214812.1 | NP_999977.1 | 1165 | 77 | ||
| GenBank mRNA | BC070025.1 | - | 1165 | 77 | ||
Set Membership: |
|