Assay Details
| Gene Symbol: rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001841.3 | NP_001001841.2 | 3 - 4 | 251 | 87 | |
| GenBank mRNA | AY163837.1 | - | 3 - 4 | 188 | 87 | |
| BC076403.1 | - | 3 - 4 | 255 | 87 | ||
More Information
Set Membership: |
|

rel
Genomic Map