Assay Details
| Gene Symbol: rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001841.3 | NP_001001841.2 | 4 - 5 | 405 | 70 | |
| GenBank mRNA | AY163837.1 | - | 4 - 5 | 342 | 70 | |
| BC076403.1 | - | 4 - 5 | 409 | 70 | ||
More Information
Set Membership: |
|

rel
Genomic Map