Assay Details
| Gene Symbol: rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001841.3 | NP_001001841.2 | 8 - 9 | 954 | 92 | |
| GenBank mRNA | AY163837.1 | - | 8 - 9 | 891 | 92 | |
| BC076403.1 | - | 8 - 9 | 958 | 92 | ||
More Information
Set Membership: |
|

rel
Genomic Map