Assay Details
| Gene Symbol: tspan9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002186.1 | NP_001002186.1 | 5 - 6 | 658 | 65 | |
| GenBank mRNA | BC074038.1 | - | 5 - 6 | 658 | 65 | |
| BC142792.1 | - | 5 - 6 | 699 | 65 | ||
More Information
Set Membership: |
|

tspan9b
Genomic Map