Assay Details
| Gene Symbol: tspan9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002186.1 | NP_001002186.1 | 6 - 7 | 788 | 63 | |
| GenBank mRNA | BC074038.1 | - | 6 - 7 | 788 | 63 | |
| BC142792.1 | - | 6 - 7 | 829 | 63 | ||
More Information
Set Membership: |
|

tspan9b
Genomic Map