Assay Details
| Gene Symbol: tjp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002366.1 | NP_001002366.1 | 7 - 8 | 1007 | 67 | |
| GenBank mRNA | BC075890.1 | - | 7 - 8 | 1007 | 67 | |
More Information
Set Membership: |
|

tjp2b
Genomic Map