Assay Details
| Gene Symbol: gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002393.1 | NP_001002393.1 | 5 - 6 | 616 | 69 | |
| GenBank mRNA | BC076194.1 | - | 5 - 6 | 616 | 69 | |
More Information
Set Membership: |
|

gpatch11
Genomic Map