Assay Details
| Gene Symbol: wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002429.1 | NP_001002429.1 | 5 - 6 | 661 | 74 | |
| GenBank mRNA | BC076138.1 | - | 5 - 6 | 661 | 74 | |
More Information
Set Membership: |
|

wdr83
Genomic Map