Assay Details
| Gene Symbol: parvaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002872.2 | NP_001002872.1 | 1 - 2 | 268 | 103 | |
| GenBank mRNA | AY648735.1 | - | 1 - 2 | 139 | 103 | |
| BC085399.1 | - | 1 - 2 | 268 | 103 | ||
| BC164207.1 | - | 1 - 2 | 173 | 103 | ||
More Information
Set Membership: |
|

parvaa
Genomic Map