Assay Details
| Gene Symbol: fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003466.1 | NP_001003466.1 | 1 - 2 | 502 | 66 | |
| XM_005173206.3 | XP_005173263.1 | 1 - 2 | 966 | 66 | ||
| XM_005173207.3 | XP_005173264.1 | 1 - 2 | 966 | 66 | ||
| GenBank mRNA | BC078364.1 | - | 1 - 2 | 502 | 66 | |
More Information
Set Membership: |
|

fam122b
Genomic Map