Assay Details
| Gene Symbol: grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003509.1 | NP_001003509.1 | 6 - 7 | 1052 | 82 | |
| GenBank mRNA | BC078350.1 | - | 6 - 7 | 1052 | 82 | |
| BC165871.1 | - | 6 - 7 | 1028 | 82 | ||
More Information
Set Membership: |
|

grwd1
Genomic Map