Assay Details
| Gene Symbol: dnajb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003571.1 | NP_001003571.1 | 2 - 3 | 1033 | 70 | |
| GenBank mRNA | BC077119.1 | - | 2 - 3 | 1072 | 70 | |
More Information
Set Membership: |
|

dnajb1a
Genomic Map