Assay Details
| Gene Symbol: sulf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003833.2 | NP_001003833.2 | 4 - 5 | 799 | 105 | |
| XM_017353509.1 | XP_017208998.1 | 4 - 5 | 809 | 105 | ||
| XM_017353510.2 | XP_017208999.1 | 6 - 7 | 1112 | 105 | ||
| XM_017353511.2 | XP_017209000.1 | 4 - 5 | 713 | 105 | ||
| XM_021469735.1 | XP_021325410.1 | 8 - 9 | 1531 | 105 | ||
| XM_021469736.1 | XP_021325411.1 | 3 - 4 | 579 | 105 | ||
| XM_021469737.1 | XP_021325412.1 | 3 - 4 | 579 | 105 | ||
| XM_021469738.1 | XP_021325413.1 | 3 - 4 | 579 | 105 | ||
| XM_021469739.1 | XP_021325414.1 | 3 - 4 | 579 | 105 | ||
| GenBank mRNA | AY332607.1 | - | 4 - 5 | 799 | 105 | |
| AY332608.1 | - | 3 - 4 | 579 | 105 | ||
More Information
Set Membership: |
|

sulf2b
Genomic Map