Assay Details
| Gene Symbol: nt5c2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004549.2 | NP_001004549.1 | 17 - 18 | 1668 | 58 | |
| XM_021479161.1 | XP_021334836.1 | 16 - 17 | 1762 | 58 | ||
| GenBank mRNA | BC081594.1 | - | 17 - 18 | 1641 | 58 | |
More Information
Set Membership: |
|

nt5c2l1
Genomic Map