Assay Details
| Gene Symbol: hat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004572.2 | NP_001004572.1 | 5 - 6 | 498 | 81 | |
| GenBank mRNA | BC081671.1 | - | 5 - 6 | 500 | 81 | |
More Information
Set Membership: |
|

hat1
Genomic Map