Assay Details
| Gene Symbol: wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309435.1 | NP_001296364.1 | 2 - 3 | 770 | 65 | |
| GenBank mRNA | AY648779.2 | - | 2 - 3 | 771 | 65 | |
| BC058071.1 | - | 1 - 2 | 60 | 65 | ||
| BC116569.1 | - | 1 - 2 | 358 | 65 | ||
More Information
Set Membership: |
|

wee1
Genomic Map