Assay Details
| Gene Symbol: jagn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005774.2 | NP_001005774.1 | 2 - 3 | 193 | 91 | |
| GenBank mRNA | BC083169.1 | - | 2 - 3 | 197 | 91 | |
More Information
Set Membership: |
|

jagn1a
Genomic Map