Assay Details
| Gene Symbol: polr3glb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005959.1 | NP_001005959.1 | 7 - 8 | 741 | 179 | |
| GenBank mRNA | BC083309.1 | - | 7 - 8 | 741 | 179 | |
More Information
Set Membership: |
|

polr3glb
Genomic Map