Assay Details
Gene Symbol: fli1rs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008780.1 | NP_001008780.1 | 6 - 7 | 841 | 129 | |
XM_005158200.3 | XP_005158257.1 | 6 - 7 | 917 | 129 | ||
XM_005158202.4 | XP_005158259.1 | 6 - 7 | 833 | 129 | ||
XM_005158203.4 | XP_005158260.1 | 6 - 7 | 830 | 129 | ||
XM_005158205.4 | XP_005158262.1 | 6 - 7 | 830 | 129 | ||
XM_009292528.3 | XP_009290803.1 | 6 - 7 | 949 | 129 | ||
XM_009292529.3 | XP_009290804.1 | 6 - 7 | 946 | 129 | ||
XM_009292530.3 | XP_009290805.1 | 6 - 7 | 948 | 129 | ||
XM_009292531.3 | XP_009290806.1 | 6 - 7 | 945 | 129 | ||
GenBank mRNA | AY839950.1 | - | 6 - 7 | 841 | 129 | |
BC055627.1 | - | 6 - 7 | 762 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
