Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 12 - 13 | 1420 | 152 | |
| XM_005158722.4 | XP_005158779.1 | 12 - 13 | 1420 | 152 | ||
| XM_005158723.4 | XP_005158780.1 | 12 - 13 | 1411 | 152 | ||
| GenBank mRNA | BC091825.1 | - | 12 - 13 | 1424 | 152 | |
| BC164273.1 | - | 11 - 12 | 1206 | 152 | ||
More Information
Set Membership: |
|

snx9a
Genomic Map