Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 15 - 16 | 1683 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snx9a| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 15 - 16 | 1683 | 96 | |
Set Membership: |
|