Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 16 - 17 | 1793 | 86 | |
| XM_005158722.4 | XP_005158779.1 | 16 - 17 | 1781 | 86 | ||
| XM_005158723.4 | XP_005158780.1 | 16 - 17 | 1772 | 86 | ||
| GenBank mRNA | BC091825.1 | - | 16 - 17 | 1785 | 86 | |
| BC164273.1 | - | 15 - 16 | 1567 | 86 | ||
More Information
Set Membership: |
|

snx9a
Genomic Map