Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 17 - 18 | 1892 | 65 | |
| XM_005158722.4 | XP_005158779.1 | 17 - 18 | 1880 | 65 | ||
| XM_005158723.4 | XP_005158780.1 | 17 - 18 | 1871 | 65 | ||
| GenBank mRNA | BC091825.1 | - | 17 - 18 | 1884 | 65 | |
| BC164273.1 | - | 16 - 17 | 1666 | 65 | ||
More Information
Set Membership: |
|

snx9a
Genomic Map