Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 5 - 6 | 610 | 99 | |
| XM_005158722.4 | XP_005158779.1 | 5 - 6 | 610 | 99 | ||
| XM_005158723.4 | XP_005158780.1 | 5 - 6 | 601 | 99 | ||
| GenBank mRNA | BC091825.1 | - | 5 - 6 | 614 | 99 | |
| BC164273.1 | - | 4 - 5 | 396 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005158723.4 transcripts.
Set Membership: |
|

snx9a
Genomic Map