Assay Details
| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 9 - 10 | 1087 | 104 | |
| XM_005158722.4 | XP_005158779.1 | 9 - 10 | 1087 | 104 | ||
| XM_005158723.4 | XP_005158780.1 | 9 - 10 | 1078 | 104 | ||
| GenBank mRNA | BC091825.1 | - | 9 - 10 | 1091 | 104 | |
| BC164273.1 | - | 8 - 9 | 873 | 104 | ||
More Information
Set Membership: |
|

snx9a
Genomic Map