Assay Details
| Gene Symbol: gadd45bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012386.2 | NP_001012386.1 | 3 - 4 | 535 | 76 | |
| GenBank mRNA | BC095819.1 | - | 3 - 4 | 535 | 76 | |
| BC165896.1 | - | 3 - 4 | 398 | 76 | ||
More Information
Set Membership: |
|

gadd45bb
Genomic Map