Assay Details
| Gene Symbol: kat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013309.3 | NP_001013327.2 | 6 - 7 | 584 | 103 | |
| XM_021476135.1 | XP_021331810.1 | 6 - 7 | 667 | 103 | ||
| XM_021476136.1 | XP_021331811.1 | 5 - 6 | 598 | 103 | ||
| GenBank mRNA | BC090743.1 | - | 6 - 7 | 553 | 103 | |
More Information
Set Membership: |
|

kat5b
Genomic Map