Assay Details
| Gene Symbol: ak5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017540.2 | NP_001017540.1 | 1 - 2 | 323 | 72 | |
| GenBank mRNA | BC092184.1 | - | 1 - 2 | 327 | 72 | |
More Information
Set Membership: |
|

ak5l
Genomic Map