Assay Details
| Gene Symbol: pcyt1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017634.1 | NP_001017634.1 | 5 - 6 | 578 | 73 | |
| XM_021480218.1 | XP_021335893.1 | 6 - 7 | 798 | 73 | ||
| GenBank mRNA | BC092816.1 | - | 5 - 6 | 578 | 73 | |
| BC164470.1 | - | 4 - 5 | 513 | 73 | ||
More Information
Set Membership: |
|

pcyt1aa
Genomic Map