Assay Details
| Gene Symbol: dnmt3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020476.2 | NP_001018312.2 | 15 - 16 | 3231 | 140 | |
| XM_005166992.4 | XP_005167049.1 | 16 - 17 | 3447 | 140 | ||
| XM_005166993.4 | XP_005167050.1 | 16 - 17 | 3420 | 140 | ||
| XM_005166994.4 | XP_005167051.1 | 15 - 16 | 3378 | 140 | ||
| XM_005166996.4 | XP_005167053.1 | 14 - 15 | 3156 | 140 | ||
| GenBank mRNA | AB196918.1 | - | 14 - 15 | 3162 | 140 | |
More Information
Set Membership: |
|

dnmt3ba
Genomic Map