Assay Details
| Gene Symbol: dpysl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020517.2 | NP_001018353.1 | 13 - 14 | 1784 | 73 | |
| XM_005155631.4 | XP_005155688.1 | 14 - 15 | 2140 | 73 | ||
| GenBank mRNA | AY987375.1 | - | 13 - 14 | 1775 | 73 | |
| AY997023.1 | - | 13 - 14 | 1782 | 73 | ||
| BC129137.1 | - | 13 - 14 | 1784 | 73 | ||
More Information
Set Membership: |
|

dpysl2b
Genomic Map