Assay Details
| Gene Symbol: dpysl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020517.2 | NP_001018353.1 | 4 - 5 | 643 | 61 | |
| XM_005155631.4 | XP_005155688.1 | 5 - 6 | 999 | 61 | ||
| GenBank mRNA | AY987375.1 | - | 4 - 5 | 634 | 61 | |
| AY997023.1 | - | 4 - 5 | 641 | 61 | ||
| BC129137.1 | - | 4 - 5 | 643 | 61 | ||
More Information
Set Membership: |
|

dpysl2b
Genomic Map