Assay Details
| Gene Symbol: dpysl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020517.2 | NP_001018353.1 | 5 - 6 | 693 | 80 | |
| XM_005155631.4 | XP_005155688.1 | 6 - 7 | 1049 | 80 | ||
| GenBank mRNA | AY987375.1 | - | 5 - 6 | 684 | 80 | |
| AY997023.1 | - | 5 - 6 | 691 | 80 | ||
| BC129137.1 | - | 5 - 6 | 693 | 80 | ||
More Information
Set Membership: |
|

dpysl2b
Genomic Map