Assay Details
| Gene Symbol: c1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020527.1 | NP_001018363.1 | 2 - 3 | 215 | 60 | |
| GenBank mRNA | BC095042.1 | - | 2 - 3 | 215 | 60 | |
| KJ778050.1 | - | 1 - 2 | 151 | 60 | ||
| KJ778051.1 | - | 1 - 2 | 151 | 60 | ||
| KJ778055.1 | - | 1 - 2 | 151 | 60 | ||
| KJ778056.1 | - | 1 - 2 | 151 | 60 | ||
| KJ778057.1 | - | 1 - 2 | 151 | 60 | ||
| KJ778058.1 | - | 1 - 2 | 151 | 60 | ||
More Information
Set Membership: |
|

c1qa
Genomic Map