Assay Details
| Gene Symbol: pin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020553.1 | NP_001018389.1 | 3 - 4 | 467 | 97 | |
| GenBank mRNA | BC095126.1 | - | 3 - 4 | 467 | 97 | |
| BC164671.1 | - | 3 - 4 | 259 | 97 | ||
More Information
Set Membership: |
|

pin4
Genomic Map