Assay Details
| Gene Symbol: tbl1xr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020583.1 | NP_001018419.1 | 12 - 13 | 1467 | 95 | |
| XM_005172823.4 | XP_005172880.1 | 12 - 13 | 1495 | 95 | ||
| GenBank mRNA | BC095221.1 | - | 12 - 13 | 1467 | 95 | |
More Information
Set Membership: |
|

tbl1xr1a
Genomic Map