Assay Details
| Gene Symbol: mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020682.1 | NP_001018518.1 | 1 - 2 | 75 | 81 | |
| GenBank mRNA | BC095767.1 | - | 1 - 2 | 75 | 81 | |
More Information
Set Membership: |
|

mrps24
Genomic Map