Assay Details
| Gene Symbol: sft2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020695.2 | NP_001018531.1 | 6 - 7 | 558 | 116 | |
| GenBank mRNA | BC095800.1 | - | 6 - 7 | 518 | 116 | |
| EH498746.1 | - | 6 - 7 | 576 | 116 | ||
More Information
Set Membership: |
|

sft2d2b
Genomic Map