Assay Details
| Gene Symbol: dnai4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020759.1 | NP_001018595.1 | 16 - 17 | 2355 | 99 | |
| GenBank mRNA | BC095632.1 | - | 16 - 17 | 2355 | 99 | |
More Information
Set Membership: |
|

dnai4
Genomic Map