Assay Details
| Gene Symbol: metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024392.1 | NP_001019563.1 | 9 - 10 | 959 | 70 | |
| GenBank mRNA | BC095283.1 | - | 9 - 10 | 959 | 70 | |
More Information
Set Membership: |
|

metap1d
Genomic Map