Assay Details
| Gene Symbol: macroh2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025502.1 | NP_001020673.1 | 6 - 7 | 941 | 68 | |
| GenBank mRNA | BC097115.1 | - | 6 - 7 | 941 | 68 | |
More Information
Set Membership: |
|

macroh2a2
Genomic Map