Assay Details
| Gene Symbol: zyg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040332.2 | NP_001035422.1 | 3 - 4 | 1034 | 120 | |
| GenBank mRNA | BC115195.1 | - | 3 - 4 | 1000 | 120 | |
More Information
Set Membership: |
|

zyg11
Genomic Map